Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037632.1 | NP_001032721.1 | 2 - 2 | 1359 | 76 |
NM_181374.2 | NP_852039.2 | 2 - 2 | 1239 | 76 | |
XM_006242390.2 | XP_006242452.1 | 2 - 2 | 1595 | 76 | |
XM_008765706.1 | XP_008763928.1 | 3 - 3 | 1165 | 76 | |
GenBank mRNA | AY177399.1 | - | 2 - 2 | 1141 | 76 |