Assay Design



Gene Transcripts

Gene Symbol
Cdc42ep1
Entrez Gene ID
315121
Gene Name
CDC42 effector protein (Rho GTPase binding) 1
Gene Aliases
-
Chromosome Location
Chr.7: 120067552 - 120071154 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079700.1 NP_001073168.1 1 - 2 564 100
GenBank mRNA BC128744.1 - 1 - 2 564 100

View Full Product Details