Assay Design



Gene Transcripts

Gene Symbol
Vom2r36
Entrez Gene ID
502301
Gene Name
vomeronasal 2 receptor, 36
Gene Aliases
RGD1560365
Chromosome Location
Chr.1: 65906344 - 66159639 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099483.1 NP_001092953.1 1 - 1 153 76

View Full Product Details