Assay Design
Gene Transcripts
- Gene Symbol
- MT-ND1
- Entrez Gene ID
-
26193
- Gene Name
- mitochondrially encoded NADH dehydrogenase 1
- Gene Aliases
-
ND1
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
NC_001665.ND1.0 |
- |
- |
844 |
120 |
View Full Product Details