Assay Design



Gene Transcripts

Gene Symbol
Crygc
Entrez Gene ID
24277
Gene Name
crystallin, gamma C
Gene Aliases
Cryg, Cryg3, Len
Chromosome Location
Chr.9: 71786246 - 71788281 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081660.1 NP_001075129.1 2 - 3 280 60
GenBank mRNA BG371711.1 - 1 - 2 60

View Full Product Details