Assay Design



Gene Transcripts

Gene Symbol
Vom2r80
Entrez Gene ID
502285
Gene Name
vomeronasal 2 receptor, 80
Gene Aliases
RGD1559687
Chromosome Location
Chr.1: 62500357 - 62508149 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111321.1 NP_001104791.1 - 481 94

View Full Product Details