Assay Design



Gene Transcripts

Gene Symbol
Vom2r4
Entrez Gene ID
308248
Gene Name
vomeronasal 2 receptor, 4
Gene Aliases
RGD1564110
Chromosome Location
Chr.1: 714270 - 719690 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099458.1 NP_001092928.1 - 1296 110

Gene Symbol
LOC102552844
Entrez Gene ID
102552844
Gene Name
vomeronasal type-2 receptor 26-like
Gene Aliases
-
Chromosome Location
Chr.10: 111716542 - 111722393 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008768523.1 XP_008766745.1 3 - 4 1720 110

View Full Product Details