Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019373.2 | NP_062246.1 | 2 - 3 | 347 | 81 |
GenBank mRNA | AF207821.1 | - | 2 - 3 | 324 | 81 |
BC089807.1 | - | 2 - 3 | 296 | 81 | |
FQ210093.1 | - | 2 - 3 | 346 | 81 | |
FQ219449.1 | - | 2 - 3 | 347 | 81 |