Assay Design
Gene Transcripts
- Gene Symbol
- Lta
- Entrez Gene ID
-
25008
- Gene Name
- lymphotoxin alpha
- Gene Aliases
-
Tnfb
- Chromosome Location
-
Chr.20: 4852671 - 4854677 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080769.2 |
NP_542947.1 |
3 - 4 |
343 |
86 |
View Full Product Details