Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173388.1 | NP_001166859.1 | 4 - 5 | 416 | 61 |
XM_006239436.2 | XP_006239498.1 | 3 - 4 | 403 | 61 | |
GenBank mRNA | BC098773.1 | - | 3 - 4 | 266 | 61 |
CB611569.1 | - | 4 - 5 | 433 | 61 |