Assay Design



Gene Transcripts

Gene Symbol
Cdh9
Entrez Gene ID
29163
Gene Name
cadherin 9, type 2, T1-cadherin
Gene Aliases
-
Chromosome Location
Chr.2: 66938794 - 67078478 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168630.1 NP_001162101.1 8 - 9 1513 102
XM_006232076.2 XP_006232138.1 10 - 11 1737 102

View Full Product Details