Assay Design



Gene Transcripts

Gene Symbol
Olr94
Entrez Gene ID
405908
Gene Name
olfactory receptor 94
Gene Aliases
-
Chromosome Location
Chr.1: 168470190 - 168471161 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001023.1 NP_001001023.1 1 - 1 748 96

View Full Product Details