Assay Design



Gene Transcripts

Gene Symbol
Moap1
Entrez Gene ID
299261
Gene Name
modulator of apoptosis 1
Gene Aliases
-
Chromosome Location
Chr.6: 126620597 - 126622532 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013101.1 NP_001013119.1 1 - 2 139 77
GenBank mRNA BC091310.1 - 1 - 2 139 77

View Full Product Details