Assay Design



Gene Transcripts

Gene Symbol
Btn3a2
Entrez Gene ID
294268
Gene Name
butyrophilin, subfamily 3, member A2
Gene Aliases
Btnl4, RGD1562488
Chromosome Location
Chr.20: 4240270 - 4247115 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166344.1 NP_001159816.1 - 913 73

View Full Product Details