Assay Design



Gene Transcripts

Gene Symbol
RT1-CE12
Entrez Gene ID
309600
Gene Name
RT1 class I, locus CE12
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008835.1 NP_001008835.1 - 1061 103

Gene Symbol
RT1-CE1
Entrez Gene ID
309603
Gene Name
RT1 class I, locus1
Gene Aliases
RT1-C/E1
Chromosome Location
Chr.20: 4638235 - 4792390 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008832.2 NP_001008832.1 6 - 7 115

View Full Product Details