Assay Design
Gene Transcripts
- Gene Symbol
- RT1-CE16
- Entrez Gene ID
-
414819
- Gene Name
- RT1 class I, locus CE16
- Gene Aliases
-
-
- Chromosome Location
-
-
- Gene Symbol
- LOC100364500
- Entrez Gene ID
-
100364500
- Gene Name
- RT1 class I, locus CE11-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.20: 2704214 - 2706648 on Build Rnor_6.0
- Gene Symbol
- LOC103690108
- Entrez Gene ID
-
103690108
- Gene Name
- class I histocompatibility antigen, Non-RT1.A alpha-1 chain-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.20: 3791235 - 3794372 on Build Rnor_6.0
- Gene Symbol
- RT1-A1
- Entrez Gene ID
-
24973
- Gene Name
- RT1 class Ia, locus A1
- Gene Aliases
-
RT1.A1(N), RT1A-1
- Chromosome Location
-
Chr.20: 5414464 - 5418012 on Build Rnor_6.0
- Gene Symbol
- RT1-CE13
- Entrez Gene ID
-
414790
- Gene Name
- RT1 class I, locus CE13
- Gene Aliases
-
-
- Chromosome Location
-
Chr.20|NW_007906368: 3928 - 4540 on Build Rnor_6.0
- Gene Symbol
- RT1-CE15
- Entrez Gene ID
-
414789
- Gene Name
- RT1 class I, locus CE15
- Gene Aliases
-
-
- Chromosome Location
-
-
- Gene Symbol
- RT1-CE2
- Entrez Gene ID
-
414779
- Gene Name
- RT1 class I, locus CE2
- Gene Aliases
-
RT1-C/E2
- Chromosome Location
-
-
- Gene Symbol
- RT1-Cl
- Entrez Gene ID
-
24977
- Gene Name
- RT1 class Ib, locus Cl
- Gene Aliases
-
RT1.A
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_206848.1 |
NP_996730.1 |
4 - 5 |
607 |
65 |
GenBank mRNA |
EU040040.1 |
- |
2 - 3 |
544 |
65 |
- Gene Symbol
- RT1-EC2
- Entrez Gene ID
-
24737
- Gene Name
- RT1 class Ib, locus EC2
- Gene Aliases
-
MHCRTA, RATMHCRTA, RT1-A, RT1-Aw2, RT1.Al, RT1.EC2, RT1Aw2, RTS
- Chromosome Location
-
-
- Gene Symbol
- Rt1.aa
- Entrez Gene ID
-
100188935
- Gene Name
- MHC class I RT1.Aa alpha-chain
- Gene Aliases
-
-
- Chromosome Location
-
-
View Full Product Details