Assay Design
Gene Transcripts
- Gene Symbol
- RT1-CE16
- Entrez Gene ID
-
414819
- Gene Name
- RT1 class I, locus CE16
- Gene Aliases
-
-
- Chromosome Location
-
-
- Gene Symbol
- RT1-EC2
- Entrez Gene ID
-
24737
- Gene Name
- RT1 class Ib, locus EC2
- Gene Aliases
-
MHCRTA, RATMHCRTA, RT1-A, RT1-Aw2, RT1.Al, RT1.EC2, RT1Aw2, RTS
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AJ004889.1 |
- |
4 - 5 |
951 |
86 |
M11071.1 |
- |
3 - 4 |
241 |
86 |
Y08530.1 |
- |
4 - 5 |
951 |
86 |
View Full Product Details