Assay Design



Gene Transcripts

Gene Symbol
Vom1r82
Entrez Gene ID
494235
Gene Name
vomeronasal 1 receptor 82
Gene Aliases
V1rc41
Chromosome Location
Chr.4: 88119838 - 88120734 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009515.1 NP_001009515.1 1 - 1 453 103
GenBank mRNA AY510313.1 - 1 - 1 453 103

View Full Product Details