Assay Design
Gene Transcripts
- Gene Symbol
- Ptgdrl
- Entrez Gene ID
-
63889
- Gene Name
- prostaglandin D2 receptor
- Gene Aliases
-
Ptgdr, Ptgdr2
- Chromosome Location
-
Chr.15: 19195606 - 19196508 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_022241.1 |
NP_071577.1 |
- |
1150 |
92 |
GenBank mRNA |
U92289.1 |
- |
- |
1150 |
92 |
- Gene Symbol
- Ptgdr
- Entrez Gene ID
-
498475
- Gene Name
- prostaglandin D2 receptor-like
- Gene Aliases
-
RGD1565108
- Chromosome Location
-
Chr.15: 18089077 - 18095979 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
FQ222068.1 |
- |
- |
1192 |
92 |
View Full Product Details