Assay Design



Gene Transcripts

Gene Symbol
Atp6ap1l
Entrez Gene ID
361875
Gene Name
ATPase, H+ transporting, lysosomal accessory protein 1-like
Gene Aliases
RGD1564936
Chromosome Location
Chr.2: 19781387 - 19808937 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191843.1 NP_001178772.1 1 - 2 131 68

View Full Product Details