Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191949.1 | NP_001178878.1 | 12 - 13 | 1364 | 64 |
XM_006229401.2 | XP_006229463.1 | 13 - 14 | 1555 | 64 | |
XM_006229403.2 | XP_006229465.1 | 13 - 14 | 1500 | 64 | |
XM_006229404.2 | XP_006229466.1 | 13 - 14 | 1555 | 64 | |
XM_008759547.1 | XP_008757769.1 | 13 - 14 | 3355 | 64 | |
XM_008759548.1 | XP_008757770.1 | 12 - 13 | 1431 | 64 | |
XM_008759549.1 | XP_008757771.1 | 13 - 14 | 1504 | 64 |