Assay Design



Gene Transcripts

Gene Symbol
Wdr93
Entrez Gene ID
499189
Gene Name
WD repeat domain 93
Gene Aliases
RGD1559862
Chromosome Location
Chr.1: 141478655 - 141528160 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191949.1 NP_001178878.1 3 - 4 556 67
XM_006229401.2 XP_006229463.1 4 - 5 747 67
XM_006229403.2 XP_006229465.1 4 - 5 692 67
XM_006229404.2 XP_006229466.1 4 - 5 747 67
XM_008759547.1 XP_008757769.1 4 - 5 2547 67
XM_008759548.1 XP_008757770.1 4 - 5 746 67
XM_008759549.1 XP_008757771.1 4 - 5 696 67
XM_008759550.1 XP_008757772.1 4 - 5 746 67
XM_008759551.1 XP_008757773.1 4 - 5 746 67
XM_008759552.1 XP_008757774.1 4 - 5 746 67
XM_008759553.1 XP_008757775.1 4 - 5 745 67

View Full Product Details