Assay Design



Gene Transcripts

Gene Symbol
LOC100359907
Entrez Gene ID
100359907
Gene Name
secretory calcium-binding phosphoprotein proline-glutamine rich 1-like
Gene Aliases
EO463, Scpppq1
Chromosome Location
Chr.14: 7026769 - 7035539 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195414.1 NP_001182343.1 4 - 5 262 74
GenBank mRNA HM015624.1 - 4 - 5 262 74

View Full Product Details