Assay Design



Gene Transcripts

Gene Symbol
Ceacam19
Entrez Gene ID
680640
Gene Name
carcinoembryonic antigen-related cell adhesion molecule 19
Gene Aliases
-
Chromosome Location
Chr.1: 80800951 - 80809769 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198970.1 NP_001185899.1 2 - 3 584 76

View Full Product Details