Assay Design



Gene Transcripts

Gene Symbol
Faim3
Entrez Gene ID
548326
Gene Name
Fas apoptotic inhibitory molecule 3
Gene Aliases
Toso
Chromosome Location
Chr.13: 47602702 - 47617065 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014843.2 NP_001014843.1 3 - 4 610 87
XM_006249715.2 XP_006249777.1 3 - 4 678 87
GenBank mRNA BC088197.1 - 3 - 4 610 87

View Full Product Details