Assay Design



Gene Transcripts

Gene Symbol
Pate-f
Entrez Gene ID
100913081
Gene Name
prostate and testis expressed protein F
Gene Aliases
-
Chromosome Location
Chr.8: 37080432 - 37093082 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267780.1 NP_001254709.1 1 - 2 64 84
GenBank mRNA JF412806.1 - 1 - 2 64 84

View Full Product Details