Assay Design



Gene Transcripts

Gene Symbol
Chmp4bl1
Entrez Gene ID
679886
Gene Name
chromatin modifying protein 4B-like 1
Gene Aliases
-
Chromosome Location
Chr.4: 67059939 - 67060988 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276456.1 NP_001263385.1 1 - 1 818 100

View Full Product Details