Assay Design
Gene Transcripts
- Gene Symbol
- GIP4
- Entrez Gene ID
-
851202
- Gene Name
- Gene Aliases
-
FUN21, YAL031C
- Chromosome Location
-
Chr.I: 84749 - 87031 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178176.1 |
NP_009371.1 |
1 - 1 |
2152 |
72 |
View Full Product Details