Assay Design
Gene Transcripts
- Gene Symbol
- OAF1
- Entrez Gene ID
-
851247
- Gene Name
- Gene Aliases
-
YAF1, YAL051W
- Chromosome Location
-
Chr.I: 48564 - 51707 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178195.2 |
NP_009349.3 |
1 - 1 |
3047 |
88 |
View Full Product Details