Assay Design



Gene Transcripts

Gene Symbol
CMC2
Entrez Gene ID
852220
Gene Name
Gene Aliases
YBL059C-A
Chromosome Location
Chr.II: 110125 - 110539 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184440.1 NP_031358.1 1 - 2 32 71

View Full Product Details