Assay Design
Gene Transcripts
- Gene Symbol
- MRP21
- Entrez Gene ID
-
852188
- Gene Name
- Gene Aliases
-
MRP50, YBL090W, bS21m
- Chromosome Location
-
Chr.II: 48825 - 49358 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178330.1 |
NP_009463.1 |
1 - 1 |
254 |
76 |
View Full Product Details