Assay Design
Gene Transcripts
- Gene Symbol
- COQ1
- Entrez Gene ID
-
852288
- Gene Name
- Gene Aliases
-
YBR003W
- Chromosome Location
-
Chr.II: 242809 - 244230 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178351.1 |
NP_009557.1 |
1 - 1 |
1356 |
71 |
View Full Product Details