Assay Design
Gene Transcripts
- Gene Symbol
- YRO2
- Entrez Gene ID
-
852343
- Gene Name
- Gene Aliases
-
YBR054W
- Chromosome Location
-
Chr.II: 343101 - 344135 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178402.1 |
NP_009610.1 |
1 - 1 |
855 |
67 |
View Full Product Details