Assay Design
Gene Transcripts
- Gene Symbol
- TIM12
- Entrez Gene ID
-
852388
- Gene Name
- Gene Aliases
-
MRS5, YBR091C
- Chromosome Location
-
Chr.II: 427155 - 427484 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178439.1 |
NP_009649.1 |
1 - 1 |
251 |
82 |
View Full Product Details