Assay Design
Gene Transcripts
- Gene Symbol
- PBY1
- Entrez Gene ID
-
852391
- Gene Name
- Gene Aliases
-
YBR094W
- Chromosome Location
-
Chr.II: 432036 - 434297 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178442.2 |
NP_009652.2 |
1 - 1 |
1646 |
105 |
View Full Product Details