Assay Design
Gene Transcripts
- Gene Symbol
- SIF2
- Entrez Gene ID
-
852399
- Gene Name
- Gene Aliases
-
EMB1, YBR103W
- Chromosome Location
-
Chr.II: 447709 - 449316 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178451.1 |
NP_009661.1 |
1 - 1 |
1548 |
89 |
View Full Product Details