Assay Design
Gene Transcripts
- Gene Symbol
- REI1
- Entrez Gene ID
-
852569
- Gene Name
- Gene Aliases
-
YBR267W
- Chromosome Location
-
Chr.II: 739841 - 741022 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178615.4 |
NP_009825.4 |
1 - 1 |
1055 |
85 |
View Full Product Details