Assay Design
Gene Transcripts
- Gene Symbol
- MAL33
- Entrez Gene ID
-
852600
- Gene Name
- Gene Aliases
-
YBR297W
- Chromosome Location
-
Chr.II: 800523 - 801929 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178645.3 |
NP_009856.3 |
1 - 1 |
1052 |
80 |
View Full Product Details