Assay Design
Gene Transcripts
- Gene Symbol
- LSB5
- Entrez Gene ID
-
850323
- Gene Name
- Gene Aliases
-
YCL034W
- Chromosome Location
-
Chr.III: 61658 - 62722 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178679.1 |
NP_009896.2 |
1 - 1 |
951 |
101 |
View Full Product Details