Assay Design
Gene Transcripts
- Gene Symbol
- APA1
- Entrez Gene ID
-
850307
- Gene Name
- Gene Aliases
-
DTP1, YCL050C
- Chromosome Location
-
Chr.III: 37836 - 38801 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178695.1 |
NP_009880.2 |
1 - 1 |
745 |
84 |
View Full Product Details