Assay Design
Gene Transcripts
- Gene Symbol
- RHB1
- Entrez Gene ID
-
850392
- Gene Name
- Gene Aliases
-
RSG1, YCR027C
- Chromosome Location
-
Chr.III: 167370 - 167999 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178742.1 |
NP_009956.2 |
1 - 1 |
550 |
99 |
View Full Product Details