Assay Design
Gene Transcripts
- Gene Symbol
- RSA4
- Entrez Gene ID
-
850435
- Gene Name
- Gene Aliases
-
YCR072C
- Chromosome Location
-
Chr.III: 240805 - 242352 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178782.1 |
NP_009997.2 |
1 - 1 |
1451 |
83 |
View Full Product Details