Assay Design
Gene Transcripts
- Gene Symbol
- DIA3
- Entrez Gene ID
-
851537
- Gene Name
- Gene Aliases
-
YDL024C
- Chromosome Location
-
Chr.IV: 408451 - 409857 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180083.1 |
NP_010260.1 |
1 - 1 |
1349 |
71 |
View Full Product Details