Assay Design
Gene Transcripts
- Gene Symbol
- PRP9
- Entrez Gene ID
-
851531
- Gene Name
- Gene Aliases
-
YDL030W
- Chromosome Location
-
Chr.IV: 397537 - 399129 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180089.1 |
NP_010254.1 |
1 - 1 |
1451 |
93 |
View Full Product Details