Assay Design
Gene Transcripts
- Gene Symbol
- ASM4
- Entrez Gene ID
-
851470
- Gene Name
- Gene Aliases
-
NUP59, YDL088C
- Chromosome Location
-
Chr.IV: 298417 - 300003 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180147.1 |
NP_010195.1 |
1 - 1 |
1449 |
91 |
View Full Product Details