Assay Design
Gene Transcripts
- Gene Symbol
- NUR1
- Entrez Gene ID
-
851469
- Gene Name
- Gene Aliases
-
YDL089W
- Chromosome Location
-
Chr.IV: 296820 - 298274 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180148.1 |
NP_010194.1 |
1 - 1 |
1346 |
70 |
View Full Product Details