Assay Design
Gene Transcripts
- Gene Symbol
- ATG9
- Entrez Gene ID
-
851406
- Gene Name
- Gene Aliases
-
APG9, AUT9, CVT6, CVT7, YDL149W
- Chromosome Location
-
Chr.IV: 184925 - 187918 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180209.1 |
NP_010132.1 |
1 - 1 |
2647 |
62 |
View Full Product Details