Assay Design
Gene Transcripts
- Gene Symbol
- LYS20
- Entrez Gene ID
-
851346
- Gene Name
- Gene Aliases
-
YDL182W
- Chromosome Location
-
Chr.IV: 133437 - 134723 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180242.1 |
NP_010099.1 |
1 - 1 |
46 |
83 |
View Full Product Details