Assay Design
Gene Transcripts
- Gene Symbol
- SEC31
- Entrez Gene ID
-
851332
- Gene Name
- Gene Aliases
-
WEB1, YDL195W
- Chromosome Location
-
Chr.IV: 107208 - 111029 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180255.2 |
NP_010086.2 |
1 - 1 |
3749 |
105 |
View Full Product Details