Assay Design
Gene Transcripts
- Gene Symbol
- SSS1
- Entrez Gene ID
-
851659
- Gene Name
- Gene Aliases
-
YDR086C
- Chromosome Location
-
Chr.IV: 616928 - 617170 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001180394.1 |
NP_010371.1 |
1 - 1 |
151 |
79 |
View Full Product Details